Product Details

SNP ID
rs9347683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:162728023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGAAGCTTGTTGCAGCTCTAGCC[A/C]AGGTCCTGCCCTCTTCCCGCCCCGC
Phenotype
MIM: 608427 MIM: 602544
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PACRG PubMed Links

Gene Details

Gene
PACRG
Gene Name
PARK2 coregulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080378.1 137 Intron NP_001073847.1
NM_001080379.1 137 UTR 5 NP_001073848.1
NM_152410.2 137 Intron NP_689623.2
XM_005266825.4 137 UTR 5 XP_005266882.1
XM_005266826.4 137 UTR 5 XP_005266883.1
XM_006715344.3 137 UTR 5 XP_006715407.1
XM_006715345.3 137 UTR 5 XP_006715408.1
XM_011535461.2 137 UTR 5 XP_011533763.1
XM_011535462.2 137 UTR 5 XP_011533764.1
XM_011535466.1 137 UTR 5 XP_011533768.1
XM_011535468.2 137 UTR 5 XP_011533770.1
XM_011535469.2 137 UTR 5 XP_011533771.1
XM_011535471.1 137 UTR 5 XP_011533773.1
XM_017010275.1 137 Intron XP_016865764.1
XM_017010276.1 137 UTR 5 XP_016865765.1
XM_017010277.1 137 Intron XP_016865766.1
XM_017010278.1 137 Intron XP_016865767.1
XM_017010279.1 137 Intron XP_016865768.1
XM_017010280.1 137 UTR 5 XP_016865769.1
XM_017010281.1 137 Intron XP_016865770.1
XM_017010282.1 137 Intron XP_016865771.1
Gene
PARK2
Gene Name
parkin RBR E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004562.2 137 Intron NP_004553.2
NM_013987.2 137 Intron NP_054642.2
NM_013988.2 137 Intron NP_054643.2
XM_011535863.1 137 Intron XP_011534165.1
XM_017010908.1 137 Intron XP_016866397.1
XM_017010909.1 137 Intron XP_016866398.1

View Full Product Details