Product Details

SNP ID
rs12407030
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:64205791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTATTAGCCCATCCTCTTTTTA[A/T]GTAGGATAAGGTCGCATTATATAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2U PubMed Links

Gene Details

Gene
UBE2U
Gene Name
ubiquitin conjugating enzyme E2 U (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152489.1 Intron NP_689702.1
XM_005270498.4 Intron XP_005270555.1
XM_006710379.3 Intron XP_006710442.1
XM_011540764.2 Intron XP_011539066.1
XM_017000379.1 Intron XP_016855868.1
XM_017000380.1 Intron XP_016855869.1
XM_017000381.1 Intron XP_016855870.1
XM_017000382.1 Intron XP_016855871.1
XM_017000383.1 Intron XP_016855872.1
XM_017000384.1 Intron XP_016855873.1
XM_017000385.1 Intron XP_016855874.1

View Full Product Details