Product Details

SNP ID
rs8092564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32679880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATTTCAGTGAATTCATTGTTA[C/T]GACAAATTGTTTGTGTTCTTCAGTT
Phenotype
MIM: 613772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL14 PubMed Links

Gene Details

Gene
KLHL14
Gene Name
kelch like family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020805.2 Intron NP_065856.1
XM_011526109.2 Intron XP_011524411.2

View Full Product Details