Product Details

SNP ID
rs1546259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69161922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGTGGGGAAGACTGATTTTGG[C/T]TTGCTTTCCTAGGTGGTGGTCTGCT
Phenotype
MIM: 612134
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLCE PubMed Links

Gene Details

Gene
GLCE
Gene Name
glucuronic acid epimerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324091.1 Intron NP_001311020.1
NM_001324092.1 Intron NP_001311021.1
NM_001324093.1 Intron NP_001311022.1
NM_001324094.1 Intron NP_001311023.1
NM_015554.2 Intron NP_056369.1
XM_005254298.3 Intron XP_005254355.1
XM_017022073.1 Intron XP_016877562.1
XM_017022074.1 Intron XP_016877563.1

View Full Product Details