Product Details

SNP ID
rs13061214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:157263036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGACACTATTGCCTAGCTAGTT[G/T]GTGGAGATCAAGGTCTTAAAAATGC
Phenotype
MIM: 609594
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VEPH1 PubMed Links

Gene Details

Gene
VEPH1
Gene Name
ventricular zone expressed PH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167911.1 Intron NP_001161383.1
NM_001167912.1 Intron NP_001161384.1
NM_001167915.1 Intron NP_001161387.1
NM_001167916.1 Intron NP_001161388.1
NM_001167917.1 Intron NP_001161389.1
NM_024621.2 Intron NP_078897.2
XM_011513134.2 Intron XP_011511436.1
XM_011513135.1 Intron XP_011511437.1

View Full Product Details