Product Details
- SNP ID
-
rs638769
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
46
- Location
-
Chr.1:42181359 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAGCGGGAAAAGTGTATGAACCA[C/G]TTAACACCTAAAATTTACCTTCTCT
- Phenotype
-
MIM: 616035
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXJ3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79076288] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXJ3
- Gene Name
- forkhead box J3
View Full Product Details