Product Details

SNP ID
rs4924208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:37940347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTATGCTCTGGCCACACAACTCC[C/T]TCATATAAGCCAAATCTGTGATAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMCO5A PubMed Links

Gene Details

Gene
TMCO5A
Gene Name
transmembrane and coiled-coil domains 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152453.3 Intron NP_689666.2
XM_005254168.3 Intron XP_005254225.1
XM_011521262.2 Intron XP_011519564.1
XM_011521263.1 Intron XP_011519565.1
XM_011521264.1 Intron XP_011519566.1

View Full Product Details