Product Details

SNP ID
rs1045282
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:111883850 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCGATACAAATGCACAGAAAAGTC[A/C]ACTTACATGAATAGCTATTCAAAGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf1 PubMed Links

Gene Details

Gene
C11orf1
Gene Name
chromosome 11 open reading frame 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022761.2 529 Silent Mutation TCA,TCC S141S NP_073598.1
XM_011542957.2 529 Silent Mutation TCA,TCC S157S XP_011541259.1
XM_011542958.2 529 Missense Mutation CAA,CCA Q135P XP_011541260.1
XM_017018129.1 529 Missense Mutation CAA,CCA Q119P XP_016873618.1
Gene
FDXACB1
Gene Name
ferredoxin-fold anticodon binding domain containing 1
There are no transcripts associated with this gene.

View Full Product Details