Product Details
- SNP ID
-
rs8445
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:27733744 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTCAAATCAAAGAAAAAGCACGC[C/G]TCAATATCACGCGTAGGAAAAACTA
- Phenotype
-
MIM: 602728
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113654695] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC25
- Gene Name
- coiled-coil domain containing 25
- Gene
- SCARA3
- Gene Name
- scavenger receptor class A member 3
View Full Product Details