Product Details

SNP ID
rs3745715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48203911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCCAGAAAGCTACAACTCCCA[A/G]CCGGCTGAGCGCCACCAGCGCTTTC
Phenotype
MIM: 609051
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf68 PubMed Links

Gene Details

Gene
C19orf68
Gene Name
chromosome 19 open reading frame 68
There are no transcripts associated with this gene.

Gene
CARD8
Gene Name
caspase recruitment domain family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184900.1 Intron NP_001171829.1
NM_001184901.1 Intron NP_001171830.1
NM_001184902.1 Intron NP_001171831.1
NM_001184903.1 Intron NP_001171832.1
NM_001184904.1 Intron NP_001171833.1
NM_014959.3 Intron NP_055774.2
XM_006723091.3 Intron XP_006723154.1
XM_006723092.3 Intron XP_006723155.1
XM_006723093.3 Intron XP_006723156.1
XM_006723094.3 Intron XP_006723157.1
XM_006723095.3 Intron XP_006723158.1
XM_006723096.3 Intron XP_006723159.1
XM_006723097.3 Intron XP_006723160.1
XM_006723098.3 Intron XP_006723161.1
XM_006723099.3 Intron XP_006723162.1
XM_006723101.2 Intron XP_006723164.1
XM_006723102.3 Intron XP_006723165.1
XM_006723104.3 Intron XP_006723167.1
XM_006723106.3 Intron XP_006723169.1
XM_006723107.3 Intron XP_006723170.1
XM_006723109.3 Intron XP_006723172.1
XM_006723110.3 Intron XP_006723173.1
XM_011526641.2 Intron XP_011524943.1
XM_011526642.2 Intron XP_011524944.1
XM_011526643.2 Intron XP_011524945.1
XM_011526644.2 Intron XP_011524946.1
XM_011526646.2 Intron XP_011524948.1
XM_011526647.2 Intron XP_011524949.1
XM_011526648.2 Intron XP_011524950.1
XM_011526650.2 Intron XP_011524952.1
XM_011526653.2 Intron XP_011524955.1
XM_017026482.1 Intron XP_016881971.1
XM_017026483.1 Intron XP_016881972.1
XM_017026484.1 Intron XP_016881973.1
XM_017026485.1 Intron XP_016881974.1
XM_017026486.1 Intron XP_016881975.1
XM_017026487.1 Intron XP_016881976.1
XM_017026488.1 Intron XP_016881977.1
XM_017026489.1 Intron XP_016881978.1
XM_017026490.1 Intron XP_016881979.1
XM_017026491.1 Intron XP_016881980.1
XM_017026492.1 Intron XP_016881981.1
XM_017026493.1 Intron XP_016881982.1
XM_017026494.1 Intron XP_016881983.1
XM_017026495.1 Intron XP_016881984.1
XM_017026496.1 Intron XP_016881985.1
XM_017026497.1 Intron XP_016881986.1
XM_017026498.1 Intron XP_016881987.1
XM_017026499.1 Intron XP_016881988.1

View Full Product Details