Product Details

SNP ID
rs12960229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21542677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCAATCAATAAATGTTCCCAAGG[G/T]TAAATACAGCAATAAATTCAAATCA
Phenotype
MIM: 609674
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ESCO1 PubMed Links

Gene Details

Gene
ESCO1
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052911.2 Intron NP_443143.2
XM_011525798.1 Intron XP_011524100.1
XM_011525799.2 Intron XP_011524101.1

View Full Product Details