Product Details
- SNP ID
-
rs2271408
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122235763 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GATCTAACCAGTCCCTAAGCTGAGA[C/T]GAGGTGGAGAAGTGTGGCTTACTTG
- Phenotype
-
MIM: 605219
MIM: 610034
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIABLO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77955945] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIABLO
- Gene Name
- diablo IAP-binding mitochondrial protein
There are no transcripts associated with this gene.
- Gene
- LOC101593348
- Gene Name
- uncharacterized LOC101593348
There are no transcripts associated with this gene.
- Gene
- VPS33A
- Gene Name
- VPS33A, CORVET/HOPS core subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022916.4 |
|
Intron |
|
|
NP_075067.2 |
View Full Product Details