Product Details
- SNP ID
-
rs1399715
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:109139643 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCATTCTTCTGCTAAAATGAAAAA[C/T]AGATTTATTTTCCCTTAGCAATCAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ22763
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77287247] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ22763
- Gene Name
- uncharacterized LOC401081
There are no transcripts associated with this gene.
View Full Product Details