Product Details
- SNP ID
-
rs1018242
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59728159 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTTTTTTTCTAGTGATTTTTATAG[C/G]CTTTCACTTGTTTTCCTTTATGAGG
- Phenotype
-
MIM: 611753
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
VMP1
PubMed Links
Gene Details
- Gene
- VMP1
- Gene Name
- vacuole membrane protein 1
There are no transcripts associated with this gene.
View Full Product Details