Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015206.2 | 7364 | Intron | NP_056021.1 | ||
XM_011521392.1 | 7364 | Intron | XP_011519694.1 | ||
XM_011521393.2 | 7364 | Intron | XP_011519695.1 | ||
XM_011521395.2 | 7364 | Intron | XP_011519697.1 | ||
XM_011521396.2 | 7364 | Intron | XP_011519698.1 | ||
XM_017022027.1 | 7364 | Intron | XP_016877516.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301203.1 | 7364 | Intron | NP_001288132.1 | ||
NM_007364.3 | 7364 | Intron | NP_031390.1 | ||
XM_017022040.1 | 7364 | UTR 3 | XP_016877529.1 | ||
XM_017022041.1 | 7364 | Intron | XP_016877530.1 |