Product Details

SNP ID
rs9893873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6756534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTGGAAAGGGAGCCGTGGGGC[A/G]GGGCAGGGGGGACAGAGCAGGGGGC
Phenotype
MIM: 606717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XAF1 PubMed Links

Gene Details

Gene
XAF1
Gene Name
XIAP associated factor 1
There are no transcripts associated with this gene.

View Full Product Details