Product Details
- SNP ID
-
rs703693
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:95866214 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACCAATTGGGAAAGATTACTCAC[C/T]GAACTAATGCTTTAAAAGCCACTGA
- Phenotype
-
MIM: 603541
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC38
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111986490] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC38
- Gene Name
- coiled-coil domain containing 38
- Gene
- LOC105369921
- Gene Name
- uncharacterized LOC105369921
There are no transcripts associated with this gene.
- Gene
- SNRPF
- Gene Name
- small nuclear ribonucleoprotein polypeptide F
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003095.2 |
547 |
UTR 3 |
|
|
NP_003086.1 |
View Full Product Details