Product Details

SNP ID
rs2137895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:53458824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATAAAATAGTGAACCAATAGT[A/T]TGTGAGAGATTTTGACTTACTACAT
Phenotype
MIM: 607686 MIM: 609732
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FIP1L1 PubMed Links

Gene Details

Gene
FIP1L1
Gene Name
factor interacting with PAPOLA and CPSF1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134937.1 4774 Intron NP_001128409.1
NM_001134938.1 4774 Intron NP_001128410.1
NM_030917.3 4774 Intron NP_112179.2
XM_005265769.4 4774 Intron XP_005265826.1
XM_005265773.4 4774 Intron XP_005265830.1
XM_005265774.4 4774 Intron XP_005265831.1
XM_005265778.4 4774 Intron XP_005265835.1
XM_005265779.4 4774 Intron XP_005265836.1
XM_005265781.4 4774 Intron XP_005265838.1
XM_005265782.4 4774 Intron XP_005265839.1
XM_017008662.1 4774 Intron XP_016864151.1
XM_017008663.1 4774 Intron XP_016864152.1
XM_017008664.1 4774 Intron XP_016864153.1
XM_017008665.1 4774 Intron XP_016864154.1
XM_017008666.1 4774 Intron XP_016864155.1
XM_017008667.1 4774 Intron XP_016864156.1
XM_017008668.1 4774 Intron XP_016864157.1
XM_017008669.1 4774 Intron XP_016864158.1
XM_017008670.1 4774 Intron XP_016864159.1
XM_017008671.1 4774 Intron XP_016864160.1
Gene
LNX1
Gene Name
ligand of numb-protein X 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126328.2 4774 Intron NP_001119800.1
NM_032622.2 4774 Intron NP_116011.2
XM_005265785.4 4774 UTR 3 XP_005265842.1
XM_017008776.1 4774 UTR 3 XP_016864265.1

View Full Product Details