Product Details

SNP ID
rs10003567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119320519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTAAATATCCTGCCAATTTGTG[C/T]AATATATAATGTAGACAAACTTCAT
Phenotype
MIM: 134640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FABP2 PubMed Links

Gene Details

Gene
FABP2
Gene Name
fatty acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000134.3 Intron NP_000125.2

View Full Product Details