Product Details

SNP ID
rs12948870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43012203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTCCACGCCCTTCAGGAGGAG[C/G]AGGCCAGACTCAAGATGAGGCTGTG
Phenotype
MIM: 600735 MIM: 607526 MIM: 604631
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFI35 PubMed Links

Gene Details

Gene
IFI35
Gene Name
interferon induced protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005533.4 254 Missense Mutation CAG,GAG Q16E NP_005524.2
XM_005257302.4 254 Missense Mutation CAG,GAG Q16E XP_005257359.1
XM_017024584.1 254 UTR 5 XP_016880073.1
XM_017024585.1 254 UTR 5 XP_016880074.1
Gene
RPL27
Gene Name
ribosomal protein L27
There are no transcripts associated with this gene.

Gene
VAT1
Gene Name
vesicle amine transport 1
There are no transcripts associated with this gene.

View Full Product Details