Product Details

SNP ID
rs1047852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1481560 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCCTCTCCCCCCAGCCCACTCC[C/T]GCCCAAACCCAAAATGCAGAGGAGA
Phenotype
MIM: 612034 MIM: 600487 MIM: 609346
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APC2 PubMed Links

Gene Details

Gene
APC2
Gene Name
APC2, WNT signaling pathway regulator
There are no transcripts associated with this gene.

Gene
C19orf25
Gene Name
chromosome 19 open reading frame 25
There are no transcripts associated with this gene.

Gene
PCSK4
Gene Name
proprotein convertase subtilisin/kexin type 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017573.4 2131 UTR 3 NP_060043.2
XM_005259586.1 2131 UTR 3 XP_005259643.1
XM_011528085.2 2131 UTR 3 XP_011526387.1
XM_011528086.2 2131 UTR 3 XP_011526388.1
XM_011528087.2 2131 UTR 3 XP_011526389.1
XM_011528088.2 2131 UTR 3 XP_011526390.1
XM_011528089.2 2131 UTR 3 XP_011526391.1
XM_011528090.2 2131 UTR 3 XP_011526392.1
XM_011528091.2 2131 UTR 3 XP_011526393.1
XM_011528092.2 2131 Intron XP_011526394.1
XM_011528093.2 2131 UTR 3 XP_011526395.1
XM_011528094.1 2131 UTR 3 XP_011526396.1
XM_011528095.2 2131 Intron XP_011526397.1
XM_011528096.1 2131 UTR 3 XP_011526398.1
XM_017026897.1 2131 Intron XP_016882386.1
Gene
REEP6
Gene Name
receptor accessory protein 6
There are no transcripts associated with this gene.

View Full Product Details