Product Details

SNP ID
rs12650678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55357004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGAACACCCCTAGTATACACC[A/C]ACCTCATTGCCTTTGCAGTTGTTAT
Phenotype
MIM: 611715
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRD5A3 PubMed Links

Gene Details

Gene
SRD5A3
Gene Name
steroid 5 alpha-reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024592.4 Intron NP_078868.1
XM_005265766.3 Intron XP_005265823.1
XM_005265767.3 Intron XP_005265824.1
XM_017008601.1 Intron XP_016864090.1
Gene
SRD5A3-AS1
Gene Name
SRD5A3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details