Product Details

SNP ID
rs34207078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46921337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGTGGGGAAAGCAGTGCCAGC[A/G]GCATCAAATCTGGAGTGGCCCTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC12 PubMed Links

Gene Details

Gene
CCDC12
Gene Name
coiled-coil domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277074.1 1610 Intron NP_001264003.1
NM_144716.5 1610 Intron NP_653317.2
XM_011533390.2 1610 Intron XP_011531692.1
XM_011533391.2 1610 Intron XP_011531693.1
XM_011533392.2 1610 Intron XP_011531694.1
XM_011533393.2 1610 Intron XP_011531695.1
XM_011533394.2 1610 Intron XP_011531696.1
XM_011533396.2 1610 UTR 3 XP_011531698.1

View Full Product Details