Product Details

SNP ID
rs10784424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64888569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGACAGGGTCTCACTTTTCCACG[C/G]AGGCTGAAGTGCAGTGGCACAATCA
Phenotype
MIM: 615077
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLJ41278 PubMed Links

Gene Details

Gene
FLJ41278
Gene Name
uncharacterized LOC400046
There are no transcripts associated with this gene.

Gene
TBC1D30
Gene Name
TBC1 domain family member 30
There are no transcripts associated with this gene.

View Full Product Details