Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001010896.2 | 265 | Missense Mutation | AGG,GGG | R72G | NP_001010896.2 |
NM_001177483.1 | 265 | UTR 3 | NP_001170954.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001173484.1 | 265 | Intron | NP_001166955.1 | ||
NM_006393.2 | 265 | Intron | NP_006384.1 | ||
NM_213569.2 | 265 | Intron | NP_998734.1 | ||
XM_005252342.4 | 265 | Intron | XP_005252399.1 | ||
XM_005252343.4 | 265 | Intron | XP_005252400.1 | ||
XM_005252344.4 | 265 | Intron | XP_005252401.1 | ||
XM_011519291.2 | 265 | Intron | XP_011517593.1 | ||
XM_017015467.1 | 265 | Intron | XP_016870956.1 | ||
XM_017015468.1 | 265 | Intron | XP_016870957.1 | ||
XM_017015469.1 | 265 | Intron | XP_016870958.1 |