Product Details

SNP ID
rs7220511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32153665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGTGAGACCCTGTCTCAAAAAA[A/G]AAAAGTGTAAAATATCCAGGATAGC
Phenotype
MIM: 613888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARGFXP2 PubMed Links

Gene Details

Gene
ARGFXP2
Gene Name
arginine-fifty homeobox pseudogene 2
There are no transcripts associated with this gene.

Gene
RHOT1
Gene Name
ras homolog family member T1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033566.2 Intron NP_001028738.1
NM_001033567.2 Intron NP_001028739.2
NM_001033568.2 Intron NP_001028740.1
NM_001288754.1 Intron NP_001275683.1
NM_001288755.1 Intron NP_001275684.1
NM_001288758.1 Intron NP_001275687.1
NM_018307.4 Intron NP_060777.3
XM_011524969.2 Intron XP_011523271.1
XM_011524971.2 Intron XP_011523273.2
XM_011524973.2 Intron XP_011523275.2
XM_011524975.2 Intron XP_011523277.1
XM_011524976.2 Intron XP_011523278.2
XM_017024819.1 Intron XP_016880308.1
XM_017024820.1 Intron XP_016880309.1
XM_017024821.1 Intron XP_016880310.1
XM_017024822.1 Intron XP_016880311.1
XM_017024823.1 Intron XP_016880312.1
XM_017024824.1 Intron XP_016880313.1
XM_017024825.1 Intron XP_016880314.1

View Full Product Details