Product Details

SNP ID
rs12936652
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:18270620 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACCTGTGTTCCAGTCCAAGAAACCT[G/A]GAGATCCAGCTGAATGTGAACATCC
Phenotype
MIM: 615498 MIM: 601243
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MIEF2 PubMed Links

Gene Details

Gene
MIEF2
Gene Name
mitochondrial elongation factor 2
There are no transcripts associated with this gene.

Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320759.1 6593 Intron NP_001307688.1
NM_004618.4 6593 Intron NP_004609.1
XM_011524001.2 6593 UTR 3 XP_011522303.1

View Full Product Details