Product Details

SNP ID
rs551835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:160887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTATGACAGCCTTTCAGGAATT[A/G]CACATTGCCTACAACATAAAGCCTT
Phenotype
MIM: 607274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP14 PubMed Links

Gene Details

Gene
USP14
Gene Name
ubiquitin specific peptidase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037334.1 Intron NP_001032411.1
NM_005151.3 Intron NP_005142.1

View Full Product Details