Product Details
- SNP ID
-
rs1042731
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:3066946 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTTCACAGCTGCAGCAAATCCCAA[C/T]TCGCCCCACGACACATTCGTCTGCC
- Phenotype
-
MIM: 603508
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYOM1
PubMed Links
Gene Details
- Gene
- MYOM1
- Gene Name
- myomesin 1
View Full Product Details