Product Details
- SNP ID
-
rs10933936
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108302447 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TTGCATTTTGATTGGTTCAAATTCC[A/G]TGTGGGGATTCCTTGATTCTGACAG
- Phenotype
-
MIM: 604371
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HHLA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74409632] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HHLA2
- Gene Name
- HERV-H LTR-associating 2
View Full Product Details