Product Details

SNP ID
rs17165388
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:134302343 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGCCTGATATCTGACACTACACA[A/C]AATACAGATAATATTTTCCCTGGGT
Phenotype
MIM: 608459
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDKL3 PubMed Links

Gene Details

Gene
CDKL3
Gene Name
cyclin dependent kinase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113575.1 Intron NP_001107047.1
NM_001300853.1 Intron NP_001287782.1
NM_016508.3 Intron NP_057592.2
XM_011543429.2 Intron XP_011541731.1
XM_011543439.2 Intron XP_011541741.1
XM_011543444.2 Intron XP_011541746.1
XM_011543445.2 Intron XP_011541747.1
XM_011543447.2 Intron XP_011541749.1
XM_017009524.1 Intron XP_016865013.1
XM_017009525.1 Intron XP_016865014.1
XM_017009526.1 Intron XP_016865015.1
XM_017009527.1 Intron XP_016865016.1
XM_017009528.1 Intron XP_016865017.1
XM_017009529.1 Intron XP_016865018.1
XM_017009530.1 Intron XP_016865019.1
XM_017009531.1 Intron XP_016865020.1
XM_017009532.1 Intron XP_016865021.1
XM_017009533.1 Intron XP_016865022.1
XM_017009534.1 Intron XP_016865023.1
XM_017009535.1 Intron XP_016865024.1
XM_017009536.1 Intron XP_016865025.1
XM_017009537.1 Intron XP_016865026.1
XM_017009538.1 Intron XP_016865027.1
XM_017009539.1 Intron XP_016865028.1
XM_017009540.1 Intron XP_016865029.1
XM_017009541.1 Intron XP_016865030.1
XM_017009542.1 Intron XP_016865031.1
XM_017009543.1 Intron XP_016865032.1
XM_017009544.1 Intron XP_016865033.1
XM_017009545.1 Intron XP_016865034.1
XM_017009546.1 Intron XP_016865035.1
XM_017009547.1 Intron XP_016865036.1
XM_017009548.1 Intron XP_016865037.1

View Full Product Details