Product Details

SNP ID
rs9568357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49921419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATAGCAGCATGAAAACAGACTGA[C/T]ACACACACCAAACAAGTTAATATGT
Phenotype
MIM: 607866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRYD7 PubMed Links

Gene Details

Gene
SPRYD7
Gene Name
SPRY domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127482.2 Intron NP_001120954.1
NM_020456.3 Intron NP_065189.1
XM_017020677.1 Intron XP_016876166.1

View Full Product Details