Product Details

SNP ID
rs11839292
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:49913975 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCAGCAAAACTACAATTTGGATCT[C/G]TTTCTAATTCTAACACTCATGTTTT
Phenotype
MIM: 607866
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPRYD7 PubMed Links

Gene Details

Gene
SPRYD7
Gene Name
SPRY domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127482.2 1945 UTR 3 NP_001120954.1
NM_020456.3 1945 UTR 3 NP_065189.1
XM_017020677.1 1945 Intron XP_016876166.1

View Full Product Details