Product Details

SNP ID
rs6509548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51518244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAACAACATTTATTTCTCTTAAG[C/G]CATCAAAGACATCTTTCTATTGCAT
Phenotype
MIM: 604405
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SIGLEC6 PubMed Links

Gene Details

Gene
SIGLEC6
Gene Name
sialic acid binding Ig like lectin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527533.2 3280 UTR 3 XP_011525835.1
XM_011527534.2 3280 Intron XP_011525836.1
XM_011527535.2 3280 UTR 3 XP_011525837.1
XM_011527536.2 3280 UTR 3 XP_011525838.1
XM_011527537.2 3280 Intron XP_011525839.1
XM_011527538.2 3280 UTR 3 XP_011525840.1
XM_011527539.2 3280 Intron XP_011525841.1
XM_017027511.1 3280 Intron XP_016883000.1

View Full Product Details