Product Details

SNP ID
rs9988929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10605929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTAAAGTATCTTCTGTCACAGGAT[A/G]TATCTACCAGAACAATTTCTTATTA
Phenotype
MIM: 604274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRA1P PubMed Links

Gene Details

Gene
KLRA1P
Gene Name
killer cell lectin like receptor A1, pseudogene
There are no transcripts associated with this gene.

Gene
MAGOHB
Gene Name
mago homolog B, exon junction complex core component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300739.2 768 UTR 3 NP_001287668.1
NM_001319985.1 768 UTR 3 NP_001306914.1
NM_018048.4 768 UTR 3 NP_060518.1
XM_005253402.1 768 Intron XP_005253459.1

View Full Product Details