Product Details
- SNP ID
-
rs12468379
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:223976079 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTCTATATCTTTGTTGTCTAGCAT[A/G]GTAGCCACTAGTCACATGTGGCCAG
- Phenotype
-
MIM: 611849
MIM: 177010
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL44
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs555814415] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL44
- Gene Name
- mitochondrial ribosomal protein L44
There are no transcripts associated with this gene.
- Gene
- SERPINE2
- Gene Name
- serpin family E member 2
View Full Product Details