Product Details
- SNP ID
-
rs1323098
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
50
- Location
-
Chr.1:78235081 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGAATAAACACTGTTGTTGGGCA[C/G]AAAAACTTCCCTTCTTGCTTTGTAC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MGC27382
PubMed Links
Gene Details
- Gene
- MGC27382
- Gene Name
- uncharacterized MGC27382
There are no transcripts associated with this gene.
View Full Product Details