Product Details

SNP ID
rs2742422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45691328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACCAAGGGGTTTATGAGTTAGA[A/G]TTTCCATTTTTATTCCTTCCTTTTG
Phenotype
MIM: 606569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIMD1-AS1 PubMed Links

Gene Details

Gene
LIMD1-AS1
Gene Name
LIMD1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SACM1L
Gene Name
SAC1 suppressor of actin mutations 1-like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319072.1 Intron NP_001306001.1
NM_001319073.1 Intron NP_001306002.1
NM_014016.4 Intron NP_054735.3
XM_011533500.2 Intron XP_011531802.1

View Full Product Details