Product Details

SNP ID
rs1277284
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:56989195 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGCACCATTCTAATGATTTTGGG[G/T]AAAGTAGCTATAGTTCAGCTTCCTG
Phenotype
MIM: 180661
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POLR2B PubMed Links

Gene Details

Gene
POLR2B
Gene Name
RNA polymerase II subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000938.2 Intron NP_000929.1
NM_001303268.1 Intron NP_001290197.1
NM_001303269.1 Intron NP_001290198.1

View Full Product Details