Product Details

SNP ID
rs1996409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44028614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTATTTTTGTCTACTGATAAGTC[A/G]CCCCTCCCCTTTGCAATGTAATCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF222 PubMed Links

Gene Details

Gene
ZNF222
Gene Name
zinc finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129996.1 Intron NP_001123468.1
NM_013360.2 Intron NP_037492.2
XM_005259208.3 Intron XP_005259265.1
XM_005259209.3 Intron XP_005259266.1
XM_017027237.1 Intron XP_016882726.1
XM_017027238.1 Intron XP_016882727.1

View Full Product Details