Product Details

SNP ID
rs2839488
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:42366077 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGGGGGAGATGTTGGCATGAACA[C/G]TCAAAAGCACTATTCTGAGACCCCA
Phenotype
MIM: 113710 MIM: 605511
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TFF1 PubMed Links

Gene Details

Gene
TFF1
Gene Name
trefoil factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003225.2 Intron NP_003216.1
Gene
TMPRSS3
Gene Name
transmembrane protease, serine 3
There are no transcripts associated with this gene.

View Full Product Details