Product Details
- SNP ID
-
rs351723
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:109224344 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCAGCAGATTTCCACACCCAGGCC[A/G]GGAGAATGAACACAGATGGGGTTGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100996634
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139139803] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100996634
- Gene Name
- transmembrane protein FLJ37396
View Full Product Details