Product Details

SNP ID
rs2282283
Assay Type
Validated
NCBI dbSNP Submissions
47
Location
Chr.1:157678581 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CAGGGGAGATTTGCAGACCTTTTGC[G/T]CAGCCTCACATACCCTGCAGCCCAG
Phenotype
MIM: 606510
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCRL3 PubMed Links

Gene Details

Gene
FCRL3
Gene Name
Fc receptor like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320333.1 2556 Intron NP_001307262.1
NM_052939.3 2556 UTR 3 NP_443171.2
XM_006711145.1 2556 UTR 3 XP_006711208.1

View Full Product Details