Product Details

SNP ID
rs77625174
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:227921008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGACGTGCTAGAAAAGCCTCTCA[G/T]TCCTTGCAGGTGTAGACCCCTCATA
Phenotype
MIM: 602863
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WNT9A PubMed Links

Gene Details

Gene
WNT9A
Gene Name
Wnt family member 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003395.2 2179 Intron NP_003386.1
XM_011544271.2 2179 UTR 3 XP_011542573.1

View Full Product Details