Product Details

SNP ID
rs6937
Assay Type
Functionally tested
NCBI dbSNP Submissions
66
Location
Chr.1:147155343 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TAAAAAGAGGACTCACATATTTAAT[T/C]CCCTTCAAAGTAGTCTTATCTCTCC
Phenotype
MIM: 602741
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PRKAB2 PubMed Links

Gene Details

Gene
PRKAB2
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005399.4 5262 UTR 3 NP_005390.1
XM_011509729.1 5262 UTR 3 XP_011508031.1
XM_017001697.1 5262 UTR 3 XP_016857186.1

View Full Product Details