Product Details

SNP ID
rs34226619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149738118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGAAAAAGGAAACTCTAGAGC[A/G]CTCTATATATAAAACCCTCAGCCTG
Phenotype
MIM: 616699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMMD2 PubMed Links

Gene Details

Gene
COMMD2
Gene Name
COMM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016094.3 Intron NP_057178.2

View Full Product Details