Product Details

SNP ID
rs34771873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196718306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCAACAGAGCGAGGCAAAAC[G/T]CCGTCTGAAGAAAAAAAAAAGGGGG
Phenotype
MIM: 615586 MIM: 610276
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP19 PubMed Links

Gene Details

Gene
CEP19
Gene Name
centrosomal protein 19
There are no transcripts associated with this gene.

Gene
PIGX
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166304.1 Intron NP_001159776.1
NM_017861.3 Intron NP_060331.3

View Full Product Details