Product Details

SNP ID
rs34950492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1714898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTAAACTCCTAAGAATGCACCA[C/G]AGAATAAAAGTTTACTTGAGAATAA
Phenotype
MIM: 605431
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAPK8IP3 PubMed Links

Gene Details

Gene
MAPK8IP3
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040439.1 Intron NP_001035529.1
NM_001318852.1 Intron NP_001305781.1
NM_015133.4 Intron NP_055948.2
XM_005255190.2 Intron XP_005255247.1
XM_006720869.2 Intron XP_006720932.1
XM_011522429.2 Intron XP_011520731.1
XM_011522430.2 Intron XP_011520732.1
XM_011522431.2 Intron XP_011520733.1
XM_011522432.2 Intron XP_011520734.1
XM_011522433.2 Intron XP_011520735.1
XM_017023079.1 Intron XP_016878568.1
XM_017023080.1 Intron XP_016878569.1
XM_017023081.1 Intron XP_016878570.1

View Full Product Details