Product Details

SNP ID
rs7848922
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:74510684 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGAGTACCCAGATCAAAGAAAAC[C/T]CTAAAATATAAAAGAGTGCTTTTTA
Phenotype
MIM: 601972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RORB PubMed Links

Gene Details

Gene
RORB
Gene Name
RAR related orphan receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006914.3 Intron NP_008845.2

View Full Product Details