Product Details

SNP ID
rs12880540
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:55781488 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTCATTCCCTCATTTGTTTGTAAG[G/T]TGGGACTTGTGTTACGGTCTTCAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00520 PubMed Links

Gene Details

Gene
LINC00520
Gene Name
long intergenic non-protein coding RNA 520
There are no transcripts associated with this gene.

View Full Product Details